
Desloratadine Related Compound A

Synonym(s):
8-Bromo-6,11-dihydro-11-(piperidin-4-ylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
Empirical Formula (Hill Notation):
C19H19BrN2
CAS Number:
Molecular Weight:
355.27
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2