
Cyclohexa-1,4-dienylglycine

Synonym(s):
(R)-(−)-2-(2,5-Dihydrophenyl)glycine, (R)-α-Amino-1,4-cyclohexadiene-1-acetic acid
Empirical Formula (Hill Notation):
C8H11NO2
CAS Number:
Molecular Weight:
153.18
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing