
Cortisol-9,11,12,12-d4

Synonym(s):
11β,17α,21-Trihydroxypregn-4-ene-3,20-dione-D4, 11β,17α,21-Trihydroxypregn-4-ene-3,20-dione-d4 (9,11,12,12-d4), 17-Hydroxycorticosterone-9,11,12,12-d4, 17-Hydroxycorticosterone-D4, 4-Pregnene-11β,17α,21-triol-3,20-dione-D4, 4-Pregnene-11β,17α,21-triol-3,20-dione-d4 (9,11,12,12-d4), Deuterated cortisol, Deuterated hydrocortisone, Hydrocortisone-9,11,12,12-d4, Kendall′s compound F (9,11,12,12-d4), Reichstein′s substance M (9,11,12,12-d4)
Empirical Formula (Hill Notation):
C21D4H26O5
CAS Number:
Molecular Weight:
366.48
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing