
Cefadroxil Related Compound I

Synonym(s):
(6R,7R)-7-[(R)-2-Amino-2-(4-hydroxyphenyl)acetamido]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylic acid
Empirical Formula (Hill Notation):
C16H17N3O5S
CAS Number:
Molecular Weight:
363.39
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing