
Anti-RRBP1 antibody produced in rabbit

Synonym(s):
Anti-180 kDa ribosome receptor homolog antibody produced in rabbit, Anti-ES/130-related protein antibody produced in rabbit, Anti-Ribosome receptor protein antibody produced in rabbit, Anti-Ribosome-binding protein 1 antibody produced in rabbit, Anti-DKFZp586A1420, Anti-ES130, Anti-FLJ36146, Anti-Ribosome binding protein 1 homolog 180kDa (dog)
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing