
Aminoallyl-dUTP sodium salt

Synonym(s):
5-[3-Aminoallyl]-2′-deoxyuridine 5′-triphosphate, AA-dUTP
Empirical Formula (Hill Notation):
C12H20N3O14P3 · xNa+
CAS Number:
Molecular Weight:
523.22 (free acid basis)
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing