Skip to Content
Merck
Acetyl coenzyme A lithium salt ≥93% (HPLC)

Acetyl coenzyme A lithium salt

Acetyl coenzyme A lithium salt ≥93% (HPLC)
Synonym(s):
Acetyl-S-CoA Li3
Empirical Formula (Hill Notation):
C23H38N7O17P3S · xLi+
CAS Number:
Molecular Weight:
809.57 (free acid basis)

Products

Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing