Skip to Content
Merck
Acetic acid glacial 100% for analysis EMPARTA® ACS

Acetic acid glacial 100%

Acetic acid glacial 100% for analysis EMPARTA® ACS
Synonym(s):
Acetic acid, Glacial acetic acid
Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Product Line
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing