
3-Acetyldeoxynivalenol

Synonym(s):
3α-Acetoxy-7α,15-dihydroxy-12,13-epoxytrichothec-9-en-8-one, 3α-Acetylvomitoxin, 3-AcDON
Empirical Formula (Hill Notation):
C17H22O7
CAS Number:
Molecular Weight:
338.35
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing