
2-Aminobenzothiazole

Synonym(s):
2-Benzothiazolamine, 2-Aminobenzothiazole
Empirical Formula (Hill Notation):
C7H6N2S
CAS Number:
Molecular Weight:
150.20
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing