
(1S,2R,5R)-(+)-Isomenthol

Synonym(s):
(1S,2R,5R)-2-Isopropyl-5-methylcyclohexanol
Empirical Formula (Hill Notation):
C10H20O
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing