Skip to Content
Merck
All Photos(22)

Key Documents

ZRB1629

Sigma-Aldrich

Anti-PLD2 Antibody, clone 6F13 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Choline phosphatase 2, EC:3.1.4.4, PLD1C, Phosphatidylcholine-hydrolyzing phospholipase D2, Phospholipase D2, hPLD2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

clone

recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 105.98 kDa
observed mol wt ~105 kDa

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 6F13 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Phospholipase D2. It targets an epitope within 21 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the N-terminal region of human Phospholipase D2.

Application

Quality Control Testing
Western Blotting Analysis: A 1:10,000 dilution of this antibody detected PLD2 in NIH3T3 cell lysate.Tested applicationsImmunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PLD2 in HeLa, A431, and d NIH 3T3 cells..Flow Cytometry Analysis: 1 μg from a representative lot detected PLD2 in one million NIH3T3 cells.Affinity Binding Assay: A representative lot of this antibody bound PLD2 peptide with a KD of 1.7 x 10-6 in an affinity binding assay.Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected PLD2 in human pancreas and human kidney tissue sections.Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-PLD2, clone 6F13 ZooMAb, Cat. No. ZRB1629, is a recombinant Rabbit monoclonal antibody that detects Phospholipase D2 and is tested in Affinity Binding Assay, Flow Cytometry and Immunocytochemistry, Immunohistochemistry, and Western Blotting.

Target description

Phospholipase D2 (UniProt: O14939; also known as EC:3.1.4.4, PLD 2, hPLD2, Choline phosphatase 2, PLD1C, Phosphatidylcholine-hydrolyzing phospholipase D2) is encoded by the PLD2 gene (Gene ID: 5338) in human. PLD2 is a peripheral membrane protein that displays ubiquitous expression and plays a role in cell membrane lipid reorganization and serves as a key cell signaling protein. It is activated by a wide variety of G-protein coupled receptors and receptor tyrosine kinases in response to a wide variety of agonists. It hydrolyzes the membrane phospholipid phosphatidylcholine to choline and the lipid messenger phosphatidic acid (PA). PA can act as a central lipid second messenger for various signaling pathways and participates in many physiological processes, including cell proliferation, migration, and cytoskeletal organization. PA is also involved in the activation of Ras‐GTPase by recruiting its exchange factor Sos or Rac1to the plasma membrane. PLD2 is reported to positively regulate proliferation of CD8+ T cells that play a critical role in cancer cell elimination and suppression of tumor progression. PLD2 is reported to degrade HIF-1 and thereby reduces VEGF secretion by endothelial cells that results in reduction of tumor neovascularization and growth. PLD2 has PX and PH domains at its regulatory N‐terminus that are putatively responsible for its GEF activity. It also has two conserved HKD domains are responsible for the lipase activity and conversion of PC into PA. Its catalytic region is localized within amino acids 441-788. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Bich, VNT., et al. (2018). Sci. Rep. 8; 6283; Frohman, MA. (2015). Trends Pharmacol. Sci. 36(3); 137-144; Park, MH., et al. (2015). Exp. Mol. Med. 47; 196; Gomez-Cambronero, J. (2012). J. Leukocyte Biol. 92(3); 461-467).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service