Skip to Content
Merck
All Photos(1)

Key Documents

SML3910

Sigma-Aldrich

Yoda2

≥98% (HPLC)

Synonym(s):

KC289, Potassium 4-(5-((2,6-dichlorobenzyl)thio)-1,3,4-thiadiazol-2-yl)benzoate, Yoda1 Analog KC289

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H9Cl2KN2O2S2
Molecular Weight:
435.39
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

Biochem/physiol Actions

Highly selective and potent agonist Piezo1, analog of Yoda1.



Yoda2 is a highly selective analog of Piezo1 agonist Yoda1 that exhibits improved potency, selectivity, solubility, stability, and efficacy.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gregory Parsonage et al.
British journal of pharmacology, 180(16), 2039-2063 (2022-12-03)
The protein PIEZO1 forms mechanically activated, calcium-permeable, non-selective cation channels in numerous cell types from several species. Options for pharmacological modulation are limited and so we modified a small-molecule agonist at PIEZO1 channels (Yoda1) to increase the ability to modulate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service