Skip to Content
Merck
All Photos(3)

Key Documents

AV31592

Sigma-Aldrich

Anti-AEBP1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AE binding protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

131 kDa

species reactivity

rabbit, horse, guinea pig, rat, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AEBP1(165)

General description

AEBP1 is a transcriptional repressor that negatively regulates PTEN. This transcription factor is involved in adipocyte growth and is a sex-specific risk factor for diet-induced obesity. Furthermore, AEBP1 is also known to have proinflammatory roles in macrophages.
Rabbit Anti-AEBP1 antibody recognizes human, mouse, rat, and pig AEBP1.

The previously assigned protein identifier Q14113 has been merged into Q8IUX7. Full details can be found on the UniProt database.

Immunogen

Synthetic peptide directed towards the N terminal region of human AEBP1

Application

Rabbit Anti-AEBP1 antibody can be used for immunohistochemical assays at concentration of 4-8μg/ml. It can also be used for western blot (0.2-5.0μg/ml) applications.

Biochem/physiol Actions

The adipocyte enhancer binding protein 1 (AEBP1) is a transcriptional repressor with carboxypeptidase (CP) activity. This protein binds to a regulatory sequence, adipocyte enhancer 1 (AE-1), located in the proximal promoter region of the adipose P2 (aP2) gene, which encodes the adipocyte fatty-acid binding protein. It is characterized as a member of the regulatory B-like CP family. This protein seems to be activated by a novel mechanism, whereby the direct binding of DNA enhances its protease activity. AEBP1 may play a role in differentiated vascular smooth muscle cells.

Sequence

Synthetic peptide located within the following region: VPPEKTKDKGKKGKKDKGPKVPKESLEGSPRPPKKGKEKPPKATKKPKEK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amin Majdalawieh et al.
Mediators of inflammation, 2010, 823821-823821 (2010-04-17)
NF-kappaB comprises a family of transcription factors that are critically involved in various inflammatory processes. In this paper, the role of NF-kappaB in inflammation and atherosclerosis and the regulation of the NF-kappaB signaling pathway are summarized. The structure, function, and
Lei Zhang et al.
Molecular medicine (Cambridge, Mass.), 11(1-12), 39-47 (2005-11-25)
Obesity is an important risk factor for heart disease, diabetes, and certain cancers, but the molecular basis for obesity is poorly understood. The transcriptional repressor AEBP1, which functions as a negative regulator of PTEN through a protein-protein interaction, is highly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service