Skip to Content
Merck
All Photos(1)

Key Documents

PHR1572

Supelco

Oxymetazoline Hydrochloride

certified reference material, pharmaceutical secondary standard

Synonym(s):

Oxymetazoline hydrochloride, 2-(3-Hydroxy-2,6-dimethyl-4-t-butylbenzyl)-2-imidazoline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24N2O · HCl
CAS Number:
Molecular Weight:
296.84
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. O0290000
traceable to USP 1486004

API family

oxymetazoline

CofA

current certificate can be downloaded

packaging

pkg of 10 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cl.Cc1cc(c(O)c(C)c1CC2=NCCN2)C(C)(C)C

InChI

1S/C16H24N2O.ClH/c1-10-8-13(16(3,4)5)15(19)11(2)12(10)9-14-17-6-7-18-14;/h8,19H,6-7,9H2,1-5H3,(H,17,18);1H

InChI key

BEEDODBODQVSIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Oxymetazoline hydrochloride belongs to the category of non-selective adrenergic drugs commonly used as eye and nose drops.

Application

Oxymetazoline hydrochloride may be used as a pharmaceutical secondary standard for the determination of the analyte in pharmaceutical formulations using spectrophotometric technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Oxymetazoline hydrochloride is a partial α2A-adrenoceptor agonist; agonist at 5-HT1A, 5-HT1B, and 5-HT1D serotonin receptors. It is a mixed agonist-antagonist at 5-HT1C serotonin receptors.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB5024 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric determination of oxymetazoline hydrochloride based on the oxidation reactions
Al-Sabha NT and Rasheed AB
Jordan Journal of Chemistry, 146(605), 1-9 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service