Skip to Content
Merck
All Photos(1)

Key Documents

39643

Millipore

Burkholderia cepacia Selective Supplement

suitable for microbiology

Synonym(s):

BCSA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.74

shelf life

limited shelf life, expiry date on the label

Quality Level

application(s)

microbiology

storage temp.

2-8°C

Application

Selective supplement for the isolation of Burkholderia cepacia in industrial samples

Components

Composition (g/vial):
Polymixin B Sulphate 300,000 IU
Gentamicin 5.00 mg
Vancomycin 1.25 mg

Preparation Note

Directions:
Reconstitute the vial with 6 ml of steril distilled water in aseptic conditions and add it to 500 ml of sterilized BCSA Agar base cooled to room temperature.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service