Skip to Content
Merck
All Photos(2)

Key Documents

RV2MAG-26K

Millipore

MILLIPLEX® Rat Vascular Injury Magnetic Bead Panel 2 - Toxicity Multiplex Assay

The analytes available for this multiplex kit are: Adiponectin, sE-Selectin, and sICAM-1.

Synonym(s):

Luminex® Rat Vascualr Injury Panel, Millipore Rat Vascualr Injury Panel, Rat Adiponectin Assay, Rat Vascular Injury Multiplex Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

rat

manufacturer/tradename

Milliplex®

assay range

accuracy: 67-92%
standard curve range: 0.1-40 ng/mL
(sICAM-1)

standard curve range: 0.1-50 ng/mL
(sE-Selectin)

standard curve range: 0.3-250 ng/mL
(Adiponectin)

standard curve range: 0.7-500 ng/mL
(von Willebrand Factor (vWF))

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

Vascular injury is important to address during the preclinical safety study of drugs. A lack of well-defined mechanisms of drug-induced vascular injury (DIVI) in animals, and the absence of specific, qualified biomarkers have become significant barriers in the development of new therapeutic treatments. In the past, hemodynamics, such as heart rate and mean arterial blood pressure, were used to measure DIVI risks but these only targeted single biomarkers. Today, some of the known vascular injury mechanisms are related to vascular wall shear and/or circumferential “hoop” stress, drug/chemical toxicity, and immunological mediation. With these known biomechanics, primary targets (smooth muscle, endothelial and immune-mediated cells) yield a number of promising candidate biomarkers for DIVI study. We provide valuable research assays to investigate multiple biomarkers of vascular injury in rat serum and plasma samples using the Luminex® xMAP® instrument platform.

The MILLIPLEX® Rat Vascular Injury Bead Panel 2 contains all the components necessary to measure the following four biomarkers in any combination using Luminex® xMAP® technology: Adiponectin, sE-Selectin, sICAM-1, von Willebrand Factor (vWF). The kit uses a 96-well format, contains a lyophilized standard cocktail, two quality controls and can measure up to 38 serum or plasma samples in duplicate.

Panel Type: Toxicity

Specificity

There was no or negligible cross-reactivity between the antibodies for an analyte and any of the other analytes within a panel.

Application

  • Analytes: Adiponectin, sE-Selectin, sICAM-1, von Willebrand Factor (vWF)
  • Recommended Sample type: serum and plasma
  • Recommended Sample dilution: 1:80
  • Assay Run Time: Overnight
  • Research Category: Toxicity

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Packaging

Everything you need in a single kit.

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Please contact Technical Service for linearity of dilution.
Sensitivity: See kit protocol for individual analytes sensitivities.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geoffrey P Dobson et al.
Annals of medicine and surgery (2012), 71, 102970-102970 (2021-11-09)
The trauma of surgery is a neglected area of research. Our aim was to examine the differential expression of genes of stress, metabolism and inflammation in the major organs of a rat following a laparotomy. Anaesthetised Sprague-Dawley rats were randomised
Makiko Ohshima et al.
Scientific reports, 6, 39377-39377 (2016-12-21)
Severe intrauterine ischemia is detrimental to the developing brain. The impact of mild intrauterine hypoperfusion on neurological development, however, is still unclear. We induced mild intrauterine hypoperfusion in rats on embryonic day 17 via arterial stenosis with metal microcoils wrapped
Fabrice Antigny et al.
Circulation, 133(14), 1371-1385 (2016-02-26)
Mutations in the KCNK3 gene have been identified in some patients suffering from heritable pulmonary arterial hypertension (PAH). KCNK3 encodes an outward rectifier K(+) channel, and each identified mutation leads to a loss of function. However, the pathophysiological role of

Related Content

MILLIPLEX® toxicity assays advance liver, kidney, and genotoxicity research by simultaneously measuring multiple toxicity biomarkers.

MILLIPLEX® toxicity assays advance liver, kidney, and genotoxicity research by simultaneously measuring multiple toxicity biomarkers.

MILLIPLEX® toxicity assays advance liver, kidney, and genotoxicity research by simultaneously measuring multiple toxicity biomarkers.

MILLIPLEX® toxicity assays advance liver, kidney, and genotoxicity research by simultaneously measuring multiple toxicity biomarkers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service