Skip to Content
Merck
All Photos(2)

Key Documents

MHSTCMAG-70KPMX

Millipore

MILLIPLEX® Mouse High Sensitivity T Cell Premixed Panel - Immunology Multiplex Assay

Simultaneous analyze low levels of cytokine and chemokine biomarker with the High Sensitivity Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.

Synonym(s):

Luminex® Mouse Immunology Panel, Millipore Mouse Immunology Panel, Mouse Immunology Protein Multiplex Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

Quality Level

species reactivity

mouse

manufacturer/tradename

Milliplex®

assay range

accuracy: 82-102%
sensitivity: 0.26-11.14 pg/mL
standard curve range: 0.10-400 pg/mL
(IL-4)

standard curve range: 0.49-2,000 pg/mL
(IFNγ)

standard curve range: 0.49-2,000 pg/mL
(IL-17A)

standard curve range: 0.49-2,000 pg/mL
(IL-7)

standard curve range: 0.49-2,000 pg/mL
(TNFα)

standard curve range: 0.73-3,000 pg/mL
(KC/GROα/CXCL1)

standard curve range: 0.98-4,000 pg/mL
(IL-2)

standard curve range: 0.98-4,000 pg/mL
(IL-5)

standard curve range: 0.98-4,000 pg/mL
(IL-6)

standard curve range: 1.34-5,500 pg/mL
(IL-10)

standard curve range: 2.93-12,000 pg/mL
(IL-12 (p70))

standard curve range: 3.17-13,000 pg/mL
(MCP-1/CCL2)

standard curve range: 3.42-14,000 pg/mL
(IL-13)

standard curve range: 3.91-16,000 pg/mL
(IL-1α)

standard curve range: 3.91-16,000 pg/mL
(IL-1β)

standard curve range: 6.1-25,000 pg/mL
(GM-CSF)

standard curve range: 7.32-30,000 pg/mL
(MIP-2/CXCL2)

standard curve range: 9.77-40,000 pg/mL
(LIX)

inter-assay cv: <15%
intra-assay cv: <10%

technique(s)

multiplexing: suitable

compatibility

configured for Premixed

detection method

fluorometric (Luminex xMAP)

shipped in

ambient

General description

Cytokines are immunomodulatory polypeptides that play key roles in both adaptive and innate immune responses. A generic term, “cytokines” includes myokines (produced by muscle cells), lymphokines (produced by activated Th cells), interleukins (acting as mediators between T cells), and chemokines (responsible for T-cell migration). One of the regulatory mechanisms of the immune system, cytokines act at the recognition, activation, or effector phases of an immune response, modulating the development and functional activities of the subtypes of T cells, B cells, and myeloid cells. Research involving cytokines plays a significant role in achieving a deeper understanding of the immune system and its multi-faceted response to most antigens, especially those responses that make up the inflammatory process.

Low levels of inflammation are involved in many clinical and sub-clinical disease states, such as autoimmune disease, cardiovascular disease, diabetes, neurological disorders, and cancer. Measuring picogram levels of cytokines, therefore, is critical for understanding the pathogenesis of these diseases.

Panel Type: Cytokines/Chemokines

Specificity

Cross Reactivty
There is no detectable cross-reactivity within the panel.
Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

  • Analytes: GM-CSF, IFNγ, IL-1α, IL-1β, IL-2, IL-4, IL-5, IL-6, IL-7, IL-10, IL-12 (p70), IL-13, IL-17A, KC/CXCL1, LIX, MCP-1, MIP-2, TNF-α
  • Recommended Sample type: serum, plasma, tissue culture or other media
  • Recommended Sample dilution: neat or 1:2 diluted serum or plasma
  • Assay Run Time: Overnight
  • Research Category: Inflammation & Immunology
  • Research Subcategory: Immune Response Metabolic Disorders Chronic Inflammatory Disease Cardiovascular Disease

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Please contact Technical Service for linearity of dilution.
Sensitivity: Please see protocol for sensitivity of individual analytes.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Fen Lin et al.
Neurobiology of stress, 15, 100341-100341 (2021-06-08)
When an individual is under stress, the undesired effect on the brain often exceeds expectations. Additionally, when stress persists for a long time, it can trigger serious health problems, particularly depression. Recent studies have revealed that depressed patients have a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service