Skip to Content
Merck
All Photos(3)

Key Documents

ABN308

Sigma-Aldrich

Anti-DISC1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Disrupted in schizophrenia 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DISC1(27185)

General description

Disrupted in schizophrenia 1 (DISC1) is a candidate gene for susceptibility to schizophrenia. It was discovered through chromosomal analysis of a large Scottish family whose members exhibited schizophrenia and related psychiatric disorders. Through yeast two-hybrid screening, it was discovered that DISC1 interacts with many members of the centrosome and cytoskeletal system including MAP1A and Nudel. More recently, DISC1 has been found to regulate the transport of a complex containing Nudel, the lissencephaly-1 (LIS1) protein, and 14-3-3epsilon from neuronal cell bodies to the axons by the action of the microtubule-dependent directed motor protein kinesin-1, also known as KIF5A. Decreased expression of DISC1 in neurons caused an accelerated rate of neuronal integration, resulting in aberrant morphological development, suggesting that DISC1 plays a role in dendritic development and synapse formation. DISC1 has at least four known isoforms.

Immunogen

KLH-conjugated linear peptide corresponding to human DISC1.

Application

Anti-DISC1 detects levels of DISC1 proteins & has been published & validated for use in IHC, ICC & WB.
Immunohistochemistry Analysis: A 1:2,000 dilution of this antibody lot detected DISC1 in rat cerebellum tissue.

Immunocytochemistry Analysis: A 1:2,000 dilution from a representative lot detected DISC1 in HeLa cells.

Western Blot Analysis: 2 µg/mL from a representative lot detected DISC1 in 10 µg/mL of SK-N-SH cell lysate and mouse brain tissue lysate.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Immunohistochemistry in human brain tissue.

Immunohistochemistry Analysis: A 1:2,000 dilution of this antibody lot detected DISC1 in human brain tissue.

Target description

~94 kDa observed. Uniprot describes 8 isoforms ranging between ~39 kDa and ~94 kDa; however, there are many additional isoforms of DISC1 known to exist. (Ishizuka, K., et al. (2007). Molecular Psychiatry. 12:897-899.)

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service