Skip to Content
Merck
All Photos(1)

Documents

157317

Sigma-Aldrich

Cyclopropyltriphenylphosphonium bromide

98%

Synonym(s):

NSC 110599

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H5P(C6H5)3Br
CAS Number:
Molecular Weight:
383.26
Beilstein:
4170278
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

178-181 °C (lit.)

SMILES string

[Br-].C1CC1[P+](c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C21H20P.BrH/c1-4-10-18(11-5-1)22(21-16-17-21,19-12-6-2-7-13-19)20-14-8-3-9-15-20;/h1-15,21H,16-17H2;1H/q+1;/p-1

InChI key

XMPWFKHMCNRJCL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Used for electrolytic polarization of Zn

Studied for potential antitumor activity

Reactant for synthesis of:
  • Keto halides for sequential azidation and intramolecular Schmidt reactions
  • Cycloporpylidenemethylarylethynynes
  • Dipeptides containing azabicyclo[3.1.0]hexane

Reactant for hydroalkynylation
Wittig reagent

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service