HPLC Separation of an Oligonucleotide and Impurities on Proteomix® SAX-NP5 versus Porous SAX
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Proteomix SAX-NP5, 15 cm x 4.6 mm I.D., 5 μm particles (Z777228)
mobile phase
(A) 0.025 M Tris buffer pH9; (B) A + 1.0 M LiCi
gradient
30 to 75% B in 45 min
flow rate
1.0 mL/min
detector
UV, 260 nm
injection
50 μL
sample
50-mer Oligo
Description
Legal Information
Proteomix is a registered trademark of Sepax Technologies