Merck
US EPA Method 608.1/608.2: GC Analysis of Organochlorine Pesticides on Equity-1701
US EPA Method 608.1/608.2: GC Analysis of Organochlorine Pesticides on Equity-1701 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SPB®-608 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
SPB®-608 Capillary GC Column
L × I.D. 30 m × 0.53 mm, df 0.50
Equity-1701 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
Equity-1701 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 1.00 μm
Equity-1701 Capillary GC Column
L × I.D. 30 m × 0.53 mm, df 0.50 μm
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 1.00 μm
SLB®-5ms Capillary GC Column
L × I.D. 15 m × 0.10 mm, df 0.10 μm
accessory
Product No.
Description
Pricing
Wheaton wide-mouth bottle
capacity 32 oz (1000 mL), amber, white polypropylene cap, PTFE flat liner
guard column
Product No.
Description
Pricing
Non-Polar Fused Silica Tubing
L × I.D. 5.0 m × 0.10 mm
Non-Polar Fused Silica Tubing
L × I.D. 5.0 m × 0.25 mm
Non-Polar Fused Silica Tubing
L × I.D. 5.0 m × 0.53 mm
recommended
Product No.
Description
Pricing
Polyimide Sealing Resin
pkg of 5 g
required but not provided
Product No.
Description
Pricing
"Y" GlasSeal Connector, Borosilicate Glass
pkg of 1 ea
"Y" GlasSeal Connector, Fused Silica
pkg of 1 ea
"Y" GlasSeal Connector, Fused Silica
pkg of 3 ea
CONDITIONS
column
Equity-1701, 30 m x 0.25 mm I.D., 0.25 μm (28372-U)
oven
75 °C (2 min), 15 °C/min to 280 °C (5 min)
inj. temp.
250 °C
detector
ECD, 300 °C
carrier gas
helium, 1.2 mL/min. constant
injection
1 μL, splitless (splitter open after 0.75 min)
liner
4 mm I.D., single taper
sample
mixture of 608.1 and 608.2 calibration solutions (40351-U and 40352-U) diluted to 100 ppb in n-hexane
Description
Legal Information
null