GC Analysis of Hydrocarbons in Gasoline on Equity-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
![GC Analysis of Hydrocarbons in Gasoline on Equity-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis suitable for GC](/deepweb/assets/sigmaaldrich/product/images/365/879/830f3256-f566-4f7b-9f56-6b5fba1a6308/800/830f3256-f566-4f7b-9f56-6b5fba1a6308.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Equity-1, 15 m x 0.10 mm I.D., 0.10 μm (28039-U)
oven
40 °C (1 min), 45 °C/min to 150 °C (2 min)
inj. temp.
175 °C
detector
FID, 175 °C
carrier gas
hydrogen, 45 cm/sec constant
injection
0.1 μL 300:1 split
liner
2 mm I.D., straight
sample
Unleaded gasoline (refinery standard), neat
Description
Legal Information
null