Merck
GC Analysis of Camphor Enantiomers on Astec® CHIRALDEX G-DP
GC Analysis of Camphor Enantiomers on Astec® CHIRALDEX™ G-DP suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX G-DP Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
standard
Product No.
Description
Pricing
(−)-Camphor
analytical standard
(±)-Camphor
purum, synthetic, ≥95.0% (GC)
(±)-Camphor
meets analytical specification of Ph. Eur., BP, racemic, ≥95% (GC)
Camphor
96%
(1S)-(−)-Camphor
95%
D-Camphor
≥97%, FG
(±)-Camphor
≥95.5%
(1R)-(+)-Camphor
98%
(±)-Camphor
SAJ first grade, ≥96.0%
CONDITIONS
column
Astec CHIRALDEX G-DP, 30 m x 0.25 mm I.D., 0.12 μm (78033AST)
oven
100 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
sample
peaks 1 & 2: camphor enantiomers
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC