GC Analysis of 1-Phenylethylamine Enantiomers (N-Acetyl Derivatives) on Astec® CHIRALDEX™ B-PM
![GC Analysis of 1-Phenylethylamine Enantiomers (N-Acetyl Derivatives) on Astec® CHIRALDEX™ B-PM suitable for GC](/deepweb/assets/sigmaaldrich/product/images/289/138/814188f2-ae49-4739-af3b-15f88e652155/800/814188f2-ae49-4739-af3b-15f88e652155.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Astec CHIRALDEX B-PM, 30 m x 0.25 mm I.D., 0.12 μm (76023AST)
oven
130 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
sample
3 mg/mL in methylene chloride
peak 1: S(-)-1-phenylethylamine (N-TFA derivative)
peak 2: R(+)-1-phenylethylamine (N-TFA derivative)
Description
Analysis Note
Can reverse elution order by using the N-TFA derivative on CHIRALDEX G-PN or B-DM columns.
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC