GC Analysis of Spearmint Essential Oil on SLB®-IL59
![GC Analysis of Spearmint Essential Oil on SLB®-IL59 suitable for GC](/deepweb/assets/sigmaaldrich/product/images/122/467/32ed468f-2718-41f8-9a0b-6097c984713e/800/32ed468f-2718-41f8-9a0b-6097c984713e.jpg)
Materials
analytical column
CONDITIONS
column
SLB-IL59, 30 m x 0.25 mm I.D., 0.2 μm (28891-U)
oven
50 °C, 3 °C/min to 300 °C
detector
FID
carrier gas
helium, 30 cm/sec, constant
Description
Analysis Note
The volatile fraction of essential oils represents a quite complex matrix, being composed of a large variety of compounds, mainly terpenes, sesquiterpenes, alcohols and other oxygenated compounds, that cannot be all separeted in a single GC analysis. The ionic liquid GC column, SLB®-IL59 showed a good resolvling power toward all the classes of compounds present in the essential oils. It also provided good peak shape for both terpenes and oxygenated compounds thus proving it is a valuable polar GC column for the analyis of these types of samples. More than 50 compounds have been identified on this column by using mass spectra databases and by co-injection of standards.
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany