Merck
  • High glucose enhances the metastatic potential of tongue squamous cell carcinoma via the PKM2 pathway.
High glucose enhances the metastatic potential of tongue squamous cell carcinoma via the PKM2 pathway.
Oncotarget (2018-01-18)
Wei Wang, Qianting He, Wangxiang Yan, Jingjing Sun, Zujian Chen, Zhonghua Liu, Zhiyuan Lu, Jinsong Hou, Yisen Shao, Xiaofeng Zhou, Anxun Wang
ABSTRACT
Previous evidence has indicated an increased cancer risk in individuals with diabetes mellitus (DM). The aim of this study was to investigate the relationship between DM (high glucose) and tongue squamous cell carcinoma (TSCC) and how high glucose mediated the metastatic potential of TSCC. The relationship between DM and TSCC was assessed in a retrospective study. The role and its mechanism of high glucose on the proliferation, metastatic potential of TSCC were investigated
Sigma-Aldrich
MISSION® esiRNA, targeting human PKM