Merck
Plasma biomarkers for neuronal ceroid lipofuscinosis.
The FEBS journal (2015-11-14)
Samantha L Hersrud, Ryan D Geraets, Krystal L Weber, Chun-Hung Chan, David A Pearce
ABSTRACT
The neuronal ceroid lipofuscinoses (NCLs) are a group of neurodegenerative genetic diseases that primarily affect children and have no known cure. A unified clinical rating scale for the juvenile form of NCL has been developed, although it has not been validated in other subtypes and does not give a true measure of the pathophysiological changes occurring during disease progression. In the present study, we have identified candidate biomarkers in blood plasma of NCL disease using multiple proteomic approaches, with the aim of developing a panel of biomarkers that could serve as a metric for therapeutic response. Candidate biomarkers were identified as proteins with levels that significantly differed between patients and controls in both sample sets. The seven candidates identified have previously been associated with neurodegenerative and inflammatory diseases. Multiplex immunoassay based testing was the most efficient and effective evaluation technique and could be employed on a broad scale to track patient response to treatment.
Sigma-Aldrich
Monoclonal Anti-APOA2 antibody produced in mouse, clone 1H6, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Apolipoprotein J Antibody, Chemicon®, from goat