Merck
  • Immuno-pharmacological characterization of group II metabotropic glutamate receptors controlling glutamate exocytosis in mouse cortex and spinal cord.
Immuno-pharmacological characterization of group II metabotropic glutamate receptors controlling glutamate exocytosis in mouse cortex and spinal cord.
British journal of pharmacology (2017-10-03)
Guendalina Olivero, Tommaso Bonfiglio, Matteo Vergassola, Cesare Usai, Barbara Riozzi, Giuseppe Battaglia, Ferdinando Nicoletti, Anna Pittaluga
ABSTRACT
We recently proposed the existence of mGlu We studied the effect of LY566332, an mGlu Cortical synaptosomes possess LY566332-sensitive autoreceptors that are slightly, although significantly, susceptible to LY2389575. In contrast, LY566332-insensitive and LY2389575-sensitive autoreceptors are present in spinal cord terminals. BINA and ML337 mimicked LY566332 and LY2389575, respectively, in controlling LY379268-mediated inhibition of glutamate exocytosis from both cortical and spinal cord synaptosomes. Incubation of cortical synaptosomes with anti-mGlu We confirmed that mGlu