Merck
  • Requisite role of vasohibin-2 in spontaneous gastric cancer formation and accumulation of cancer-associated fibroblasts.
Requisite role of vasohibin-2 in spontaneous gastric cancer formation and accumulation of cancer-associated fibroblasts.
Cancer science (2017-09-30)
Yasuhiro Suzuki, Shuji Kitahara, Takuya Suematsu, Masanobu Oshima, Yasufumi Sato
ABSTRACT
The vasohibin (VASH) family consists of two genes, VASH1 and VASH2. VASH1 is mainly expressed in vascular endothelial cells and suppresses angiogenesis in an autocrine manner, whereas VASH2 is mainly expressed in cancer cells and exhibits pro-angiogenic activity. Employing adenomatous polyposis coli gene mutant mice, we recently reported on the role of Vash2 in the spontaneous formation of intestinal tumors. In this study, we used K19-Wnt1/C2mE (Gan) mice and examined the role of Vash2 in spontaneous gastric cancer formation. Gan mice spontaneously develop gastric tumors by activation of Wnt and prostaglandin E2 signaling pathways in gastric mucosa after 30 weeks of age. Expression of Vash2 mRNA was significantly increased in gastric tumor tissues compared with normal stomach tissues. When Gan mice were crossed with the Vash2-deficient (Vash2
Sigma-Aldrich
MISSION® esiRNA, targeting human VASH2 (1)