Merck
  • Single-molecule visualization of conformational changes and substrate transport in the vitamin B
Single-molecule visualization of conformational changes and substrate transport in the vitamin B
Nature communications (2017-11-23)
Joris M H Goudsmits, Dirk Jan Slotboom, Antoine M van Oijen
ABSTRACT
ATP-binding cassette (ABC) transporters form the largest class of active membrane transport proteins. Binding and hydrolysis of ATP by their highly conserved nucleotide-binding domains drive conformational changes of the complex that mediate transport of substrate across the membrane. The vitamin B
Sigma-Aldrich
Phospho(enol)pyruvic acid trisodium salt hydrate, ≥97% (enzymatic)
Sigma-Aldrich
Triton X-100, laboratory grade