Merck
  • The role of apelin in the healing of water-immersion and restraint stress-induced gastric damage.
The role of apelin in the healing of water-immersion and restraint stress-induced gastric damage.
The journal of physiological sciences : JPS (2016-07-03)
İlknur Birsen, Burcu Gemici, Nuray Acar, İsmail Üstünel, V Nimet İzgüt-Uysal
ABSTRACT
The objective of this study was to explore the role of apelin in the healing of gastric lesions induced by stress. Male Wistar rats were exposed to water immersion and restraint stress (WIRS) for 6 h with or without the apelin receptor antagonist F13A. The rats were killed on the 1st, 3rd, 5th or 10th day after the end of stress induction. Apelin and hypoxia-inducible factor-1α expression was increased on the 1st day after the end of stress exposure and was decreased daily thereafter. However, F13A retarded the healing of gastric lesions by preventing the improvement of mucosal blood flow, prostaglandin E
Sigma-Aldrich
[Ala13]-Apelin-13 trifluoroacetate salt, ≥96% (HPLC)
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder