Merck
  • Ratiometric Fluorescence Detection of Phosphorylated Amino Acids Through Excited-State Proton Transfer by Using Molecularly Imprinted Polymer (MIP) Recognition Nanolayers.
Ratiometric Fluorescence Detection of Phosphorylated Amino Acids Through Excited-State Proton Transfer by Using Molecularly Imprinted Polymer (MIP) Recognition Nanolayers.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-09-05)
Wei Wan, Ana B Descalzo, Sudhirkumar Shinde, Hardy Weißhoff, Guillermo Orellana, Börje Sellergren, Knut Rurack
ABSTRACT
A 2,3-diaminophenazine bis-urea fluorescent probe monomer (1) was developed. It responds to phenylphosphate and phosphorylated amino acids in a ratiometric fashion with enhanced fluorescence accompanied by the development of a redshifted emission band arising from an excited-state proton transfer (ESPT) process in the hydrogen-bonded probe/analyte complex. The two urea groups of 1 form a cleft-like binding pocket (K
Sigma-Aldrich
2-tert-Butyl-4-methylphenol, 99%
Sigma-Aldrich
Benzyl methacrylate, 96%, contains monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
2,3-Diaminophenazine, 90%
Sigma-Aldrich
2-Isocyanatoethyl methacrylate, contains ≤0.1% BHT as inhibitor, 98%