Merck
  • Isolation of six novel 7-oxo- or urso-type secondary bile acid-producing bacteria from rat cecal contents.
Isolation of six novel 7-oxo- or urso-type secondary bile acid-producing bacteria from rat cecal contents.
Journal of bioscience and bioengineering (2017-07-29)
Sarinya Tawthep, Satoru Fukiya, Ja-Young Lee, Masahito Hagio, Yoshitoshi Ogura, Tetsuya Hayashi, Atsushi Yokota
ABSTRACT
Understanding the dynamics of secondary bile acid (SBA) formation in the gut by SBA-producing bacteria is important for host health, as SBAs have been shown to affect host pathophysiology and gut microbiota composition. However, our knowledge of SBA producers is limited in light of the diversity of gut microbes. Here, we isolated six novel SBA-producing bacteria from rat cecal contents, all of which were members of known species of gut microbes. Anaerostipes caccae D10, Bacteroides nordii C5, Clostridioides difficile D7, and Clostridium cadaveris G11 were capable of oxidizing cholic acid and chenodeoxycholic acid into 7-oxo-derivatives with varying yields. B. nordii C5 and its type strain JCM 12987
Sigma-Aldrich
Sodium chenodeoxycholate, ≥97%