Merck
  • PDE3 inhibition by C-type natriuretic peptide-induced cGMP enhances cAMP-mediated signaling in both non-failing and failing hearts.
PDE3 inhibition by C-type natriuretic peptide-induced cGMP enhances cAMP-mediated signaling in both non-failing and failing hearts.
European journal of pharmacology (2017-07-13)
Silja Meier, Kjetil Wessel Andressen, Jan Magnus Aronsen, Ivar Sjaastad, Karina Hougen, Tor Skomedal, Jan-Bjørn Osnes, Eirik Qvigstad, Finn Olav Levy, Lise Román Moltzau
ABSTRACT
We have previously shown that the natriuretic peptide receptor B (NPR-B) agonist C-type natriuretic peptide (CNP) enhances cyclic adenosine 3´,5´-monophosphate (cAMP)-mediated signaling in failing hearts, through cyclic guanosine 3´,5´-monophosphate (cGMP)-mediated phosphodiesterase (PDE) 3 inhibition. As several signaling pathways are importantly changed in failing hearts, it could not be taken for granted that this crosstalk would be the same in non-failing hearts. Thus, we wanted to clarify to which extent this effect of CNP occurred also in non-failing hearts. Inotropic and lusitropic responses were measured in muscle strips and cGMP levels, localized cAMP levels, cAMP-PDE activity and mRNA levels were analyzed in isolated cardiomyocytes from left ventricles of non-failing and failing rat hearts. CNP increased cGMP and enhanced β
Sigma-Aldrich
Guanosine 3′,5′-cyclic monophosphate, ≥98% (HPLC), powder
Sigma-Aldrich
Natriuretic Peptide, C-Type, ≥95% (HPLC)
Sigma-Aldrich
Benzamidine hydrochloride, 99%
Sigma-Aldrich
(+)-Biotin 4-nitrophenyl ester, 98%
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
Rolipram, solid, ≥98% (HPLC)
Sigma-Aldrich
Cilostamide, phosphodiesterase inhibitor
Sigma-Aldrich
EHNA hydrochloride, ≥98% (HPLC)
Adrenaline tartrate, European Pharmacopoeia (EP) Reference Standard