- The PPARα activator fenofibrate inhibits voltage-dependent K
The PPARα activator fenofibrate inhibits voltage-dependent K
European journal of pharmacology (2017-07-19)
Hongliang Li, Sung Eun Shin, Mi Seon Seo, Jin Ryeol An, Won-Kyo Jung, Kwon-Soo Ha, Eun-Taek Han, Seok-Ho Hong, Hyoweon Bang, Young Min Bae, Amy L Firth, Il-Whan Choi, Won Sun Park
PMID28716724
ABSTRACT
We examined the effects of the PPARα activator fenofibrate on voltage-dependent K
MATERIALS