Merck
  • Role of Macromolecular Crowding on Stability and Iron Release Kinetics of Serum Transferrin.
Role of Macromolecular Crowding on Stability and Iron Release Kinetics of Serum Transferrin.
The journal of physical chemistry. B (2017-08-25)
Sandeep Kumar, Deepak Sharma, Rajesh Kumar
ABSTRACT
The macromolecular crowding influences the structural stability and functional properties of transferrin (Tf). The equilibrium as well as kinetic studies of Tf at different concentrations of crowding agents (dextran 40, dextran 70, and ficoll 70) and at a fixed concentration of dextran 40 under different concentrations of NaCl at pH 7.4 and 5.6 (±1) revealed that (i) the crowder environment increases the diferric-Tf (Fe