Merck
  • Xanthohumol, a hop-derived prenylated flavonoid, promotes macrophage reverse cholesterol transport.
Xanthohumol, a hop-derived prenylated flavonoid, promotes macrophage reverse cholesterol transport.
The Journal of nutritional biochemistry (2017-05-16)
Hiroshi Hirata, Harumi Uto-Kondo, Masatsune Ogura, Makoto Ayaori, Kazusa Shiotani, Ami Ota, Youichi Tsuchiya, Katsunori Ikewaki
ABSTRACT
Xanthohumol, a prominent prenyl flavonoid from the hop plant (Humulus lupulus L.), is suggested to be antiatherogenic since it reportedly increases high-density lipoprotein (HDL) cholesterol levels. It is not clear whether xanthohumol promotes reverse cholesterol transport (RCT), the most important antiatherogenic property of HDL; therefore, we investigated the effects of xanthohumol on macrophage-to-feces RCT using a hamster model as a CETP-expressing species. In vivo RCT experiments showed that xanthohumol significantly increased fecal appearance of the tracer derived from intraperitoneally injected [
Sigma-Aldrich
Sandoz 58-035, >98% (HPLC), powder