Merck
  • Application of visible-light photosensitization to form alkyl-radical-derived thin films on gold.
Application of visible-light photosensitization to form alkyl-radical-derived thin films on gold.
Beilstein journal of nanotechnology (2017-10-20)
Rashanique D Quarels, Xianglin Zhai, Neepa Kuruppu, Jenny K Hedlund, Ashley A Ellsworth, Amy V Walker, Jayne C Garno, Justin R Ragains
ABSTRACT
Visible-light irradiation of phthalimide esters in the presence of the photosensitizer [Ru(bpy)
Sigma-Aldrich
DCC, 99%
Sigma-Aldrich
Undecanoic acid, 98%