Merck
  • Establishing a green platform for biodiesel synthesis via strategic utilization of biochar and dimethyl carbonate.
Establishing a green platform for biodiesel synthesis via strategic utilization of biochar and dimethyl carbonate.
Bioresource technology (2017-06-13)
Jechan Lee, Jong-Min Jung, Jeong-Ik Oh, Yong Sik Ok, Eilhann E Kwon
ABSTRACT
To establish a green platform for biodiesel production, this study mainly investigates pseudo-catalytic (non-catalytic) transesterification of olive oil. To this end, biochar from agricultural waste (maize residue) and dimethyl carbonate (DMC) as an acyl acceptor were used for pseudo-catalytic transesterification reaction. Reaction parameters (temperature and molar ratio of DMC to olive oil) were also optimized. The biodiesel yield reached up to 95.4% under the optimal operational conditions (380°C and molar ratio of DMC to olive oil (36:1)). The new sustainable environmentally benign biodiesel production introduced in this study is greener and faster than conventional transesterification reactions.
Product Description
Supelco 37 Component FAME Mix, certified reference material, TraceCERT®, in dichloromethane (varied conc.), ampule of 1 mL