Merck
  • Development and validation of a reversed-phase HPLC method for analysis of radiochemical purity in [
Development and validation of a reversed-phase HPLC method for analysis of radiochemical purity in [
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2017-05-20)
Shiou-Shiow Farn, Yuen-Han Yeh, Chang-Chin Li, Ching-Shiuann Yang, Jenn-Tzong Chen, Chung-Shan Yu, Wuu-Jyh Lin
ABSTRACT
[
Sigma-Aldrich
3,3-Dimethylglutaric acid, 98%