Merck
  • π-Extended triptycene-based material for capillary gas chromatographic separations.
π-Extended triptycene-based material for capillary gas chromatographic separations.
Analytica chimica acta (2017-09-17)
Yinhui Yang, Qinsi Wang, Meiling Qi, Xuebin Huang
ABSTRACT
Triptycene-based materials feature favorable physicochemical properties and unique molecular recognition ability that offer good potential as stationary phases for capillary gas chromatography (GC). Herein, we report the investigation of utilizing a π-extended triptycene material (denoted as TQPP) for GC separations. As a result, the TQPP capillary column exhibited high column efficiency of 4030 plates m
Sigma-Aldrich
2,2,3-Trimethylbutane, ≥99%
Sigma-Aldrich
o-Cymene, 98%
Sigma-Aldrich
3-Methylheptane, 98%