Merck
  • Application of magnetic OMS-2 in sequencing batch reactor for treating dye wastewater as a modulator of microbial community.
Application of magnetic OMS-2 in sequencing batch reactor for treating dye wastewater as a modulator of microbial community.
Journal of hazardous materials (2017-07-18)
Fei Pan, Yang Yu, Aihua Xu, Dongsheng Xia, Youmin Sun, Zhengqing Cai, Wen Liu, Jie Fu
ABSTRACT
The potential and mechanism of synthesized magnetic octahedral molecular sieve (Fe
Sigma-Aldrich
Manganese(II) acetate, 98%