Merck
  • Changes in pro-inflammatory markers and leucine concentrations in response to Nordic Walking training combined with vitamin D supplementation in elderly women.
Changes in pro-inflammatory markers and leucine concentrations in response to Nordic Walking training combined with vitamin D supplementation in elderly women.
Biogerontology (2017-03-21)
A Gmiat, J Mieszkowski, K Prusik, K Prusik, J Kortas, A Kochanowicz, A Radulska, M Lipiński, M Tomczyk, J Jaworska, J Antosiewicz, E Ziemann
ABSTRACT
Mechanisms underpinning age-related decreases in muscle strength and muscle mass relate to chronic inflammation. Physical activity induces an anti-inflammatory effect, but it is modulated by additional factors. We hypothesized that vitamin D, which has also anti-inflammatory activity will modify adaptation to exercise and reduce inflammation in elderly women. Twenty-seven women aged 67 ± 8 years were included and divided into groups with baseline vitamin D concentration more than 20 ng mL
Sigma-Aldrich
Perfluoropentanoic acid, 97%