Merck
The Detection and Characterization of QoI-Resistant
Frontiers in plant science (2017-07-18)
Ayodeji S Owati, Bright Agindotan, Julie S Pasche, Mary Burrows
ABSTRACT
Ascochyta blight (AB) of pulse crops (chickpea, field pea, and lentils) causes yield loss in Montana, where 1.2 million acres was planted to pulses in 2016. Pyraclostrobin and azoxystrobin, quinone outside inhibitor (QoI) fungicides, have been the choice of farmers for the management of AB in pulses. However, a G143A mutation in the
Sigma-Aldrich
Salicylhydroxamic acid, 99%