- Assembling Porphyrins into Extended Network Structures by Employing Aromatic Dicarboxylates: Synthesis, Metal Exchange, and Heterogeneous Catalytic Studies.
Assembling Porphyrins into Extended Network Structures by Employing Aromatic Dicarboxylates: Synthesis, Metal Exchange, and Heterogeneous Catalytic Studies.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-04-20)
Gargi Dutta, Ajay Kumar Jana, Srinivasan Natarajan
PMID28422333
ABSTRACT
Three new metal-organic porphyrinic framework compounds of zinc and 5,10,15,20-tetrakis(4-pyridyl)porphyrin (TPyP) have been synthesized under solvothermal conditions. The compounds [Zn
MATERIALS