Merck
  • Rosuvastatin and Atorvastatin Are Ligands of the Human Constitutive Androstane Receptor/Retinoid X Receptor
Rosuvastatin and Atorvastatin Are Ligands of the Human Constitutive Androstane Receptor/Retinoid X Receptor
Drug metabolism and disposition: the biological fate of chemicals (2017-05-26)
Tadeja Režen, Mateja Hafner, Sandhya Kortagere, Sean Ekins, Vesna Hodnik, Damjana Rozman
ABSTRACT
Statins are well known lipid lowering agents that inhibit the enzyme 3-hydroxy-3-methylglutaryl-CoA (HMG-CoA) reductase. They also activate drug metabolism but their exact receptor-mediated action has not been proven so far. We tested whether atorvastatin and rosuvastatin are direct ligands of human constitutive androstane receptor (CAR). We measured binding activities of atorvastatin and rosuvastatin to the human constitutive androstane receptor/retinoid X receptor
Sigma-Aldrich
CITCO, ≥98% (HPLC), solid