Merck
  • Aripiprazole and Riluzole treatment alters behavior and neurometabolites in young ADHD rats: a longitudinal 1H-NMR spectroscopy study at 11.7T.
Aripiprazole and Riluzole treatment alters behavior and neurometabolites in young ADHD rats: a longitudinal 1H-NMR spectroscopy study at 11.7T.
Translational psychiatry (2017-08-02)
F Rizzo, A Abaei, E Nespoli, J M Fegert, B Hengerer, V Rasche, T M Boeckers
ABSTRACT
Attention deficit hyperactivity disorder (ADHD), Tourette syndrome (TS) as well as obsessive compulsive disorder (OCD) are co-occurring neurodevelopmental diseases that share alterations of frontocortical neurometabolites. In this longitudinal study we investigated the behavioral and neurochemical effects of aripiprazole and riluzole treatment in juvenile spontaneously hypertensive rats (SHR), a model for ADHD. For neurochemical analysis we employed in vivo magnetic resonance spectroscopy (MRS). Spectra from voxels located at the central striatum and prefrontal cortex were acquired postnatally from day 35 to 50. In the SHR strain only, treatments reduced repetitive grooming and climbing behavior. The absolute quantification of cerebral metabolites in vivo using localized 1H-MRS at 11.7T showed significant alterations in SHR rats compared to controls (including glutamine, aspartate and total NAA). In addition, drug treatment reduced the majority of the detected metabolites (glutamate and glutamine) in the SHR brain. Our results indicate that the drug treatments might influence the hypothesized 'hyperactive' state of the cortico-striatal-thalamo-cortical circuitries of the SHR strain. Furthermore, we could show that behavioral changes correlate with brain region-specific alterations in neurometabolite levels in vivo. These findings should serve as reference for animal studies and for the analysis of neurometabolites in selected human brain regions to further define neurochemical alterations in neuropsychiatric diseases.
Sigma-Aldrich
ECO TWEEN® 80, viscous liquid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
220,00 US$
50 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
391,00 US$
Sigma-Aldrich
TWEEN® 80, viscous liquid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
220,00 US$
50 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
391,00 US$