Merck
  • Neuronal nitric oxide synthase-derived hydrogen peroxide effect in grafts used in human coronary bypass surgery.
Neuronal nitric oxide synthase-derived hydrogen peroxide effect in grafts used in human coronary bypass surgery.
Clinical science (London, England : 1979) (2017-04-01)
Patrick Wander Endlich, Rosária Dias Aires, Roberta Lins Gonçalves, Eduardo Damasceno Costa, Janaína de Paula Arantes Ângelo, Lucas Ferreira Alves, Rafaela Fernandes da Silva, Bruno Almeida Rezende, Steyner F Cortes, Virgínia Soares Lemos
ABSTRACT
Recently, H
Sigma-Aldrich
1-[2-(Trifluoromethyl)phenyl]imidazole
Sigma-Aldrich
Acetylcholine bromide, ≥99%