Merck
  • CMG2/ANTXR2 regulates extracellular collagen VI which accumulates in hyaline fibromatosis syndrome.
CMG2/ANTXR2 regulates extracellular collagen VI which accumulates in hyaline fibromatosis syndrome.
Nature communications (2017-06-13)
Jérôme Bürgi, Béatrice Kunz, Laurence Abrami, Julie Deuquet, Alessandra Piersigilli, Sabine Scholl-Bürgi, Ekkehart Lausch, Sheila Unger, Andrea Superti-Furga, Paolo Bonaldo, F Gisou van der Goot
ABSTRACT
Loss-of-function mutations in capillary morphogenesis gene 2 (CMG2/ANTXR2), a transmembrane surface protein, cause hyaline fibromatosis syndrome (HFS), a severe genetic disorder that is characterized by large subcutaneous nodules, gingival hypertrophy and severe painful joint contracture. Here we show that CMG2 is an important regulator of collagen VI homoeostasis. CMG2 loss of function promotes accumulation of collagen VI in patients, leading in particular to nodule formation. Similarly, collagen VI accumulates massively in uteri of Antxr2-/- mice, which do not display changes in collagen gene expression, and leads to progressive fibrosis and sterility. Crossing Antxr2-/- with Col6a1-/- mice leads to restoration of uterine structure and reversion of female infertility. We also demonstrate that CMG2 may act as a signalling receptor for collagen VI and mediates its intracellular degradation.
Sigma-Aldrich
Anti-Fibronectin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Laminin from human placenta, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Benzamidine, ≥95.0%
Sigma-Aldrich
MISSION® esiRNA, targeting human ANTXR2
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Anti-MMP-14 Antibody, catalytic domain, clone LEM-2/15.8, clone LEM-2/15.8, Chemicon®, from mouse